missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-5-phenyl-4-isoxazolecarboxylic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 203.2 |
Percent Purity | 97% |
Chemical Name or Material | 3-Methyl-5-phenyl-4-isoxazolecarboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'