missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-5-(3-nitrophenyl)cyclohex-2-enone, TRC

Product Code. 30480126
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30480126 50 mg 50mg
30465714 250 mg 250mg
30476952 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30480126 Supplier Toronto Research Chemicals Supplier No. TRCM33899850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10323-98-5
Molecular Formula C13H13NO3
Molecular Weight (g/mol) 231.25
Synonym 3-Methyl-5-(3-nitrophenyl)-2-cyclohexen-1-one, 3-Methyl-5-(m-nitrophenyl)-2-cyclohexen-1-one
IUPAC Name 3-methyl-5-(3-nitrophenyl)cyclohex-2-en-1-one
SMILES CC1=CC(=O)CC(C1)c2cccc(c2)[N+](=O)[O-]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C13H13NO3/c1-9-5-11(8-13(15)6-9)10-3-2-4-12(7-10)14(16)17/h2-4,6-7,11H,5,8H2,1H3
Formula Weight 231.0895
Chemical Name or Material 3-Methyl-5-(3-nitrophenyl)cyclohex-2-enone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.