missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-4-phenylpyrazole, TRC

Product Code. 30480462
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30480462 25 mg 25mg
30473590 100 mg 100mg
30476166 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30480462 Supplier Toronto Research Chemicals Supplier No. TRCM32589425MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13788-84-6
Molecular Formula C10 H10 N2
Molecular Weight (g/mol) 158.2
IUPAC Name 3-methyl-4-phenyl-1H-pyrazole
SMILES Cc1n[nH]cc1c2ccccc2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C10H10N2/c1-8-10(7-11-12-8)9-5-3-2-4-6-9/h2-7H,1H3,(H,11,12)
Formula Weight 158.0844
Chemical Name or Material 3-Methyl-4-phenylpyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.