missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-4-nonanone, TRC

Product Code. 30479426
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30479426 100 mg 100mg
30473527 10 mg 10mg
30469157 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30479426 Supplier Toronto Research Chemicals Supplier No. TRCM544315100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35778-39-3
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.26
IUPAC Name 3-methylnonan-4-one
SMILES CCCCCC(=O)C(C)CC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H20O/c1-4-6-7-8-10(11)9(3)5-2/h9H,4-8H2,1-3H3
Formula Weight 156.1514
Chemical Name or Material 3-Methyl-4-nonanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.