missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-4-nitrothiobenzamide, 97%, Thermo Scientific™

Catalog Number 15490818
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
886364-22-3
C8H8N2O2S
196.224
MFCD06738330
QNPFIXDGSZDAGC-UHFFFAOYSA-N
3-methyl-4-nitrothiobenzamide, 3-methyl-4-nitrobenzothioamide, 3-methyl-4-nitrobenzene-1-carbothioamide
55255791
3-methyl-4-nitrobenzenecarbothioamide
CC1=C(C=CC(=C1)C(=S)N)[N+](=O)[O-]
This item is not returnable. View return policy
886364-22-3
C8H8N2O2S
196.224
MFCD06738330
QNPFIXDGSZDAGC-UHFFFAOYSA-N
3-methyl-4-nitrothiobenzamide, 3-methyl-4-nitrobenzothioamide, 3-methyl-4-nitrobenzene-1-carbothioamide
55255791
3-methyl-4-nitrobenzenecarbothioamide
CC1=C(C=CC(=C1)C(=S)N)[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Beilstein 19566115
Formula Weight 196.22
Percent Purity 97%
Chemical Name or Material 3-Methyl-4-nitrothiobenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'