missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-4-nitrobenzonitrile, 97%

Product Code. p-7081359
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
Unit Size:
10g
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11443684

Brand: Thermo Scientific Alfa Aesar A11301.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 96784-54-2 | C8H6N2O2 | 162.148 g/mol

3-Methyl-4-nitrobenzonitrile is used to produce 4-amino-3-methyl-benzonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-4-nitrobenzonitrile is used to produce 4-amino-3-methyl-benzonitrile.

Solubility
Insoluble in water

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96784-54-2
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
MDL Number MFCD00017615
InChI Key IHVNKSXTJZNBQA-UHFFFAOYSA-N
Synonym benzonitrile, 3-methyl-4-nitro, 5-cyano-2-nitrotoluene, 3-methyl-4-nitro-benzonitrile, 3-methyl-4-nitrobenzenecarbonitrile, 4-nitro-m-tolunitrile, 4-cyano-2-methylnitrobenzene, ihvnksxtjznbqa-uhfffaoysa
PubChem CID 2801434
IUPAC Name 3-methyl-4-nitrobenzonitrile
SMILES CC1=C(C=CC(=C1)C#N)[N+](=O)[O-]

Specifications

Melting Point 78°C to 84°C
Quantity 1 g
UN Number UN3439
Beilstein 2443778
Solubility Information Insoluble in water
Formula Weight 162.15
Percent Purity 97%
Chemical Name or Material 3-Methyl-4-nitrobenzonitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.