missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-4-nitroanisole, 97%, Thermo Scientific™

Product Code. 11436066
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11436066 100 g 100g
11416066 5 g 5g
11426066 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11436066 Supplier Thermo Scientific Alfa Aesar Supplier No. B22348.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5367-32-8
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00007167
InChI Key RTZOGYCMIMOVHU-UHFFFAOYSA-N
Synonym 3-methyl-4-nitroanisole, 5-methoxy-2-nitrotoluene, benzene, 4-methoxy-2-methyl-1-nitro, 1-methoxy-3-methyl-4-nitrobenzene, 4-methoxy-2-methyl-1-nitro-benzene, anisole, 3-methyl-4-nitro, 3-methoxy-6-nitrotoluene, 2-nitro-5-methoxy toluene, 2-nitro-5-methoxymethylbenzene, pubchem4149
PubChem CID 79330
IUPAC Name 4-methoxy-2-methyl-1-nitrobenzene
SMILES CC1=C(C=CC(=C1)OC)[N+](=O)[O-]

Specifications

Melting Point 48°C to 50°C
Flash Point >110°C (230°F)
Quantity 100 g
Beilstein 2328600
Formula Weight 167.16
Percent Purity 97%
Chemical Name or Material 3-Methyl-4-nitroanisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.