missing translation for 'onlineSavingsMsg'
Learn More

3-methyl-4-isoxazolecarboxylic acid, 97%, Thermo Scientific™

Product Code. 10443963
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10443963 1 g 1g
10433963 250 mg 250mg
10452644 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10443963 Supplier Thermo Scientific Maybridge Supplier No. MO01252DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 17153-20-7
Molecular Formula C5H5NO3
Molecular Weight (g/mol) 127.099
InChI Key YBLSBWHFPXDRHC-UHFFFAOYSA-N
Synonym 3-methylisoxazole-4-carboxylic acid, 3-methyl-4-isoxazolecarboxylic acid, 3-methyl-4-isoxazolecarboxylicacid, 3-methyl-isoxazole-4-carboxylic acid, 4-isoxazolecarboxylic acid, 3-methyl, 3-methylisoxazole-4-carboxylicacid, pubchem10465, acmc-1c78i, 3-methylisoxazole-s-carboxylic acid, 4-isoxazolecarboxylicacid,3-methyl
PubChem CID 4589692
IUPAC Name 3-methyl-1,2-oxazole-4-carboxylic acid
SMILES CC1=NOC=C1C(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 127.1
Percent Purity 97%
Chemical Name or Material 3-Methyl-4-isoxazolecarboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.