missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-3-trimethylsiloxy-1-butyne, 97%

Product Code. 11354968
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354968 50 g 50g
11374968 2 g 2g
11364968 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11354968 Supplier Thermo Scientific Alfa Aesar Supplier No. 043922.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17869-77-1 | C8H16OSi | 156.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17869-77-1
Molecular Formula C8H16OSi
Molecular Weight (g/mol) 156.3
MDL Number MFCD00053867
InChI Key JNRUXZIXAXHXTN-UHFFFAOYSA-N
Synonym 3-methyl-3-trimethylsilyloxy-1-butyne, trimethyl 2-methylbut-3-yn-2-yloxy silane, trimethyl 2-methylbut-3-yn-2-yl oxy silane, 1,1-dimethyl-2-propynyl oxy trimethylsilane, 3-methyl-3-trimethylsiloxy-1-butyne, acmc-1brai, silane, 1,1-dimethyl-2-propynyl oxy trimethyl, 3-trimethylsiloxy-3-methyl-1-butyne
PubChem CID 87344
IUPAC Name trimethyl(2-methylbut-3-yn-2-yloxy)silane
SMILES CC(C)(C#C)O[Si](C)(C)C

Specifications

Boiling Point 52°C (75 mmHg)
Odor Characteristic
Refractive Index 1.4052
Quantity 50 g
UN Number UN1993
Formula Weight 156.3
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Methyl-3-trimethylsiloxy-1-butyne

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.