missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-3-oxetanemethanol, 98%

Product Code. 10031754 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10031754 50 g Glass bottle 50g
10451104 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10031754 Supplier Thermo Scientific Acros Supplier No. 240840500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3143-02-0 | C5H10O2 | 102.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3143-02-0
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00010273
InChI Key NLQMSBJFLQPLIJ-UHFFFAOYSA-N
Synonym 3-methyl-3-oxetanemethanol, 3-methyloxetan-3-yl methanol, 3-hydroxymethyl-3-methyloxetane, 3-oxetanemethanol, 3-methyl, 3-methyl-3-oxetanyl methanol, 3-methyloxetane-3-methanol, 3-methyl-3-oxethanemethanol, 3-methyl-3-hydroxymethyloxetane, 3-hydroxymethyl-3-methyl-oxetane, 3-methyl-oxetan-3-yl-methanol
PubChem CID 137837
IUPAC Name (3-methyloxetan-3-yl)methanol
SMILES CC1(COC1)CO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0340g/mL
Boiling Point 122.0°C (70.0 mmHg)
Flash Point 97°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 50 g
Specific Gravity 1.034
Formula Weight 102.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Methyl-3-oxetanemethanol
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methyl-3-oxetanemethanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.