missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-3-hexanol, 99%

Product Code. 11381637
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11381637 25 g 25g
11371637 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11381637 Supplier Thermo Scientific Alfa Aesar Supplier No. L04625.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 597-96-6 | C7H16O | 116.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 597-96-6
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.204
MDL Number MFCD00021839
InChI Key KYWJZCSJMOILIZ-UHFFFAOYSA-N
Synonym 3-methyl-3-hexanol, 3-hexanol, 3-methyl, 2-ethyl-2-pentanol, 3-methyl-hexanol-3, 3-methyl-hexanol-3 german, acmc-20an0r, 4-01-00-01749 beilstein handbook reference
PubChem CID 11708
IUPAC Name 3-methylhexan-3-ol
SMILES CCCC(C)(CC)O

Specifications

Density 0.823
Boiling Point 143°C
Flash Point 45°C (113°F)
Refractive Index 1.4235
Quantity 25 g
UN Number UN1987
Beilstein 1719032
Formula Weight 116.2
Percent Purity 99%
Chemical Name or Material 3-Methyl-3-hexanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.