missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-3-heptanol, 98%

Product Code. p-7025475
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400027 1 g 1g
11410027 5 g 5g
11420027 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11400027 Supplier Thermo Scientific Alfa Aesar Supplier No. B20959.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5582-82-1 | C8H18O | 130.231 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5582-82-1
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.231
MDL Number MFCD00021841
InChI Key PQOSNJHBSNZITJ-UHFFFAOYSA-N
Synonym 3-methyl-3-heptanol, 3-heptanol, 3-methyl, 2-ethyl-2-hexanol, butylethylmethylcarbinol, 3-heptanol, 3-methyl-8ci 9ci, acmc-209lpn
PubChem CID 11710
IUPAC Name 3-methylheptan-3-ol
SMILES CCCCC(C)(CC)O

Specifications

Density 0.83
Boiling Point 64°C (201 mmHg)
Flash Point 58°C (136°F)
Quantity 1 g
UN Number UN1987
Formula Weight 130.23
Percent Purity 98%
Chemical Name or Material 3-Methyl-3-heptanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.