missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-3-buten-1-ol, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 0.85 |
| Boiling Point | 130°C to 133°C |
| Flash Point | 36°C (97°F) |
| Refractive Index | 1.433 |
| Quantity | 100 mL |
| UN Number | UN1987 |
| Beilstein | 1071239 |
| Formula Weight | 86.13 |
| Percent Purity | 97% |
| Chemical Name or Material | 3-Methyl-3-buten-1-ol |
Safety and Handling
Product Identifier
- 3-Methyl-3-buten-1-ol
- Warning
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contain: 3-Methyl-3-buten-1-ol
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?