missing translation for 'onlineSavingsMsg'
Learn More

(3-Methyl-2-thienyl)methylamine, 97%, Maybridge

Product Code. 10089561
Change view
Click to view available options
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10089561 Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10089561 Supplier Thermo Scientific Maybridge Supplier No. SP01443DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 104163-35-1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C6H9NS
MDL Number MFCD02093988
Quantity 1 g
Synonym 3-methylthiophen-2-yl methanamine, 3-methyl-2-thienyl methylamine, 3-methylthiophene-2-methylamine, 2-thiophenemethanamine,3-methyl, 2-thiophenemethanamine, 3-methyl, acmc-20apo3, 3-metylthiophene-2-metylamine, 2-aminomethyl-3-methylthiophene, 3-methylthiophene-2-methanamine, 3-methylthien-2-yl methylamine
InChI Key SWZNXCABBUKIPZ-UHFFFAOYSA-N
SMILES CC1=C(SC=C1)CN
IUPAC Name (3-methylthiophen-2-yl)methanamine
Molecular Weight (g/mol) 127.205
PubChem CID 2798756
Formula Weight 127.21
Chemical Name or Material (3-Methyl-2-thienyl)methylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.