missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-pyrazolin-5-one, 98+%

Product Code. 11433963
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433963 50 g 50g
11423963 10 g 10g
11443963 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11433963 Supplier Thermo Scientific Alfa Aesar Supplier No. A10253.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 108-26-9 | C4H6N2O | 98.105 g/mol

3-Methyl-2-pyrazolin-5-one is used in the preparation of spirocyclic [indole-3,4'-pyrano[2,3-c]pyrazole] by reacting with isatins and malonitrile in the presence of sodium bromide as electrolyte.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-2-pyrazolin-5-one is used in the preparation of spirocyclic [indole-3,4′-pyrano[2,3-c]pyrazole] by reacting with isatins and malonitrile in the presence of sodium bromide as electrolyte.

Solubility
Slightly soluble in ethanol.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-26-9
Molecular Formula C4H6N2O
Molecular Weight (g/mol) 98.105
MDL Number MFCD00020699
InChI Key NHLAPJMCARJFOG-UHFFFAOYSA-N
Synonym 3-methyl-2-pyrazolin-5-one, 3-methyl-5-pyrazolone, 3-methyl-1h-pyrazol-5 4h-one, 3h-pyrazol-3-one, 2,4-dihydro-5-methyl, 3-methyl-pyrazolon-5, 2-pyrazolin-5-one, 3-methyl, 3-methyl-4,5-dihydro-1h-pyrazol-5-one, 5-methyl-2,4-dihydro-3h-pyrazol-3-one, 2,4-dihydro-5-methyl-3h-pyrazol-3-one, 3-methyl-pyrazolon-5 german
PubChem CID 7920
IUPAC Name 3-methyl-1,4-dihydropyrazol-5-one
SMILES CC1=NNC(=O)C1

Specifications

Melting Point 220°C to 224°C
Quantity 50 g
Solubility Information Slightly soluble in ethanol.
Formula Weight 98.11
Percent Purity ≥98%
Chemical Name or Material 3-Methyl-2-pyrazolin-5-one

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.