missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-nitrobenzoic acid, 98%

Product Code. 11441707
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11441707 25 g 25g
11451707 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11441707 Supplier Thermo Scientific Alfa Aesar Supplier No. B25682.14
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 5437-38-7 | C8H7NO4 | 181.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5437-38-7
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.15
MDL Number MFCD00007180
InChI Key DGDAVTPQCQXLGU-UHFFFAOYSA-N
Synonym 3-methyl-2-nitrobenzoic acid, 2-nitro-m-toluic acid, 2-nitro-3-methylbenzoic acid, benzoic acid, 3-methyl-2-nitro, unii-61wop984ab, 3-methyl-2-nitro-benzoic acid, 3-methyl-2-nitrobenzoicacid, pubchem8352, acmc-1ashh, dsstox_cid_5640
PubChem CID 21575
IUPAC Name 3-methyl-2-nitrobenzoic acid
SMILES CC1=CC=CC(C(O)=O)=C1[N+]([O-])=O

Specifications

Melting Point 219°C to 223°C
Quantity 25 g
Formula Weight 181.15
Percent Purity 98%
Chemical Name or Material 3-Methyl-2-nitrobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.