missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-furoic acid, 98+%

Product Code. 11381397
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11381397 5 g 5g
11371397 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11381397 Supplier Thermo Scientific Alfa Aesar Supplier No. L01763.06
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4412-96-8 | C6H6O3 | 126.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4412-96-8
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.11
MDL Number MFCD00014111
InChI Key BNYIQEFWGSXIKQ-UHFFFAOYSA-N
Synonym 3-methyl-2-furoic acid, 2-furancarboxylic acid, 3-methyl, 3-methyl-2-furancarboxylic acid, unii-5jp68392hm, 3-methyl-furan-2-carboxylic acid, 3-methyl-2-furoicacid, methylpyromucic acid, 3-methyl furoic acid, 3-methylpyromucic acid, acmc-20a5im
PubChem CID 78127
IUPAC Name 3-methylfuran-2-carboxylic acid
SMILES CC1=C(OC=C1)C(O)=O

Specifications

Melting Point 133°C to 137°C
Quantity 5 g
Beilstein 116183
Formula Weight 126.11
Percent Purity ≥98%
Chemical Name or Material 3-Methyl-2-furoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.