missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-cyclopenten-1-one, 98%, stabilized

Product Code. 10317870 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10317870 50 g Glass bottle 50g
10285730 10 g Glass bottle 10g
11914841 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10317870 Supplier Thermo Scientific Acros Supplier No. 163590500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2758-18-1 | C6H8O | 96.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2758-18-1
Molecular Formula C6H8O
Molecular Weight (g/mol) 96.13
InChI Key CHCCBPDEADMNCI-UHFFFAOYSA-N
Synonym 3-methyl-2-cyclopenten-1-one, 3-methylcyclopent-2-enone, 2-cyclopenten-1-one, 3-methyl, 3-methyl-2-cyclopentenone, 1-methyl-1-cyclopenten-3-one, unii-6v7rsw7273, fema no. 3435, 3-methylcyclopentenone, acmc-1crym, 3-methyl-2cyclopenten-1-one
PubChem CID 17691
IUPAC Name 3-methylcyclopent-2-en-1-one
SMILES CC1=CC(=O)CC1

Specifications

Melting Point 3.0°C to 5.0°C
Color Brown-Yellow to Yellow
Density 0.9800g/mL
Boiling Point 74.0°C (15.0 mmHg)
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C5H5(=O)
Refractive Index 1.4870 to 1.4900
Quantity 50 g
Beilstein 07,I,46
Specific Gravity 0.98
Solubility Information Solubility in water: miscible
Stabilizer 0.1% hydroquinone
Formula Weight 96.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Methyl-2-cyclopenten-1-one
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methyl-2-cyclopenten-1-one, stabilized
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H335-May cause respiratory irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.