missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-cyclopenten-1-one, 98%, stab. with 0.1% hydroquinone

Product Code. 11429070
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429070 10 g 10g
11439070 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11429070 Supplier Thermo Scientific Alfa Aesar Supplier No. A13993.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2758-18-1 | C6H8O | 96.129 g/mol

It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

Solubility
Fully miscible in water.

Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent, heat. Store in refrigerator.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2758-18-1
Molecular Formula C6H8O
Molecular Weight (g/mol) 96.129
MDL Number MFCD00001403
InChI Key CHCCBPDEADMNCI-UHFFFAOYSA-N
Synonym 3-methyl-2-cyclopenten-1-one, 3-methylcyclopent-2-enone, 2-cyclopenten-1-one, 3-methyl, 3-methyl-2-cyclopentenone, 1-methyl-1-cyclopenten-3-one, unii-6v7rsw7273, fema no. 3435, 3-methylcyclopentenone, acmc-1crym, 3-methyl-2cyclopenten-1-one
PubChem CID 17691
IUPAC Name 3-methylcyclopent-2-en-1-one
SMILES CC1=CC(=O)CC1

Specifications

Melting Point 2°C
Density 0.98
pH 4.9
Boiling Point 72°C to 74°C (15 mmHg)
Flash Point 65°C (149°F)
Odor Pleasant
Refractive Index 1.489
Quantity 10 g
Beilstein 1280476
Solubility Information Fully miscible in water.
Formula Weight 96.13
Percent Purity 98%
Chemical Name or Material 3-Methyl-2-cyclopenten-1-one, stabilized with 0.1% hydroquinone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.