missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-buten-1-yl Thiolacetate (3-Methyl-2-buten-1-thiol precursor), TRC

Product Code. 30480626
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30480626 10 mg 10mg
30482012 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30480626 Supplier Toronto Research Chemicals Supplier No. TRCM29440510MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33049-93-3
Molecular Formula C7 H12 O S
Molecular Weight (g/mol) 144.23
IUPAC Name S-(3-methylbut-2-enyl) ethanethioate
SMILES CC(=CCSC(=O)C)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C7H12OS/c1-6(2)4-5-9-7(3)8/h4H,5H2,1-3H3
Formula Weight 144.0609
Chemical Name or Material 3-Methyl-2-buten-1-yl Thiolacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.