missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-buten-1-ol, 98+%

Product Code. 11423107
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423107 500 g 500g
11403107 25 g 25g
11413107 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423107 Supplier Thermo Scientific Alfa Aesar Supplier No. A16089.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 556-82-1 | C5H10O | 86.13 g/mol

3-Methyl-2-buten-1-ol is used as a reagent for protection of carboxylic acids as their 3-methyl-2-buten-1-yl (Prenyl) esters. It is also used in perfumery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-2-buten-1-ol is used as a reagent for protection of carboxylic acids as their 3-methyl-2-buten-1-yl (Prenyl) esters. It is also used in perfumery.

Solubility
Soluble in water (170 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 556-82-1
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00002916
InChI Key ASUAYTHWZCLXAN-UHFFFAOYSA-N
Synonym 3-methyl-2-buten-1-ol, prenol, 3,3-dimethylallyl alcohol, 2-buten-1-ol, 3-methyl, prenyl alcohol, 3-methyl-2-butenol, dimethylallyl alcohol, 3-methyl-2-butenyl alcohol, 3-methylcrotyl alcohol, 3-methyl-2-butene-1-ol
PubChem CID 11173
ChEBI CHEBI:16019
IUPAC Name 3-methylbut-2-en-1-ol
SMILES CC(C)=CCO

Specifications

Density 0.86
Boiling Point 139°C to 140°C
Flash Point 43°C (109°F)
Refractive Index 1.444
Quantity 500 g
UN Number UN1987
Beilstein 1633479
Solubility Information Soluble in water (170g/L at 20°C).
Formula Weight 86.13
Percent Purity ≥98%
Chemical Name or Material 3-Methyl-2-buten-1-ol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.