missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-butanone, 98%

Product Code. 11450358
Change view
Click to view available options
Quantity:
250 mL
1000 mL
Unit Size:
1000mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450358 250 mL 250mL
11440358 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11450358 Supplier Thermo Scientific Alfa Aesar Supplier No. B24527.AK
  • ONLINE EXCLUSIVE PRICE
    £32.80
    £29.65 / 250mL
    Save £3.15
    9% Off
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 563-80-4 | C5H10O | 86.13 g/mol

3-Methyl-2-butanone is used for synthesis of heterocyclic compounds and important intermediate of medicine. It is used as an industrial solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-2-butanone is used for synthesis of heterocyclic compounds and important intermediate of medicine. It is used as an industrial solvent.

Solubility
Soluble in water (8.21 mg/ml at 20°C), ether, alcohol, acetone, and ethanol.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 563-80-4
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00008919
InChI Key SYBYTAAJFKOIEJ-UHFFFAOYSA-N
Synonym 3-methyl-2-butanone, methyl isopropyl ketone, isopropyl methyl ketone, 2-acetylpropane, 2-butanone, 3-methyl, methyl butanone-2, mipk, 3-methylbutanone, ketone, isopropyl methyl, 2-methyl-3-butanone
PubChem CID 11251
IUPAC Name 3-methylbutan-2-one
SMILES CC(C)C(C)=O

Specifications

Melting Point -92°C
Density 0.805
Boiling Point 94°C to 95°C
Flash Point −1°C (30°F)
Odor Fruit-like
Refractive Index 1.388
Quantity 250 mL
UN Number UN2397
Beilstein 1071238
Merck Index 14,6090
Solubility Information Soluble in water (8.21mg/ml at 20°C),ether,alcohol,acetone,and ethanol.
Formula Weight 86.13
Percent Purity 98%
Chemical Name or Material 3-Methyl-2-butanone
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methyl-2-butanone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
Supplemental information
  • MIXTURE LIST-Contain: Methyl isopropyl ketone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.