missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-butanone, 98%, Thermo Scientific Chemicals

Catalog Number 10042990
Click to view available options
Quantity:
250 mL
1 L
5 L
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
563-80-4
C5H10O
86.13
MFCD00008919
SYBYTAAJFKOIEJ-UHFFFAOYSA-N
3-methyl-2-butanone, methyl isopropyl ketone, isopropyl methyl ketone, 2-acetylpropane, 2-butanone, 3-methyl, methyl butanone-2, mipk, 3-methylbutanone, ketone, isopropyl methyl, 2-methyl-3-butanone
11251
3-methylbutan-2-one
CC(C)C(C)=O
This item is not returnable. View return policy
563-80-4
C5H10O
86.13
MFCD00008919
SYBYTAAJFKOIEJ-UHFFFAOYSA-N
3-methyl-2-butanone, methyl isopropyl ketone, isopropyl methyl ketone, 2-acetylpropane, 2-butanone, 3-methyl, methyl butanone-2, mipk, 3-methylbutanone, ketone, isopropyl methyl, 2-methyl-3-butanone
11251
3-methylbutan-2-one
CC(C)C(C)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point -92.0°C
Color Colorless
Density 0.8000g/mL
Boiling Point 94.0°C to 95.0°C
Flash Point 6°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHCOCH3
Refractive Index 1.3870 to 1.3890
Quantity 1 L
Beilstein 01, 682
Merck Index 15, 6162
Specific Gravity 0.8
Solubility Information Solubility in water: 6g/L (20 c). Other solubilities: miscible with most common organic solvents
Formula Weight 86.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Methyl-2-butanone
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methyl-2-butanone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'