missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-3-Methyl-2-butanol, 98%

Product Code. 11387405
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387405 100 g 100g
11377405 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387405 Supplier Thermo Scientific Alfa Aesar Supplier No. L05347.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 598-75-4 | C5H12O | 88.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
3-Methyl-2-butanol has been used as a reference for microbially produced, volatile organic compounds (MVOC) in GC-MS analyses. It acts as a plant metabolite and polar solvent.

Solubility
Miscible in Ether, Alcohols. Slightly soluble in water. (2.8g/100mL, 30°C)

Notes
Incompatible materials are Strong oxidizing agents, Reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-75-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00004527
InChI Key MXLMTQWGSQIYOW-UHFFFAOYNA-N
Synonym 3-methyl-2-butanol, sec-isoamyl alcohol, 2-butanol, 3-methyl, 2-methyl-3-butanol, isopropylmethylcarbinol, methylisopropylcarbinol, 1,2-dimethylpropanol, 1,2-dimethyl-1-propanol, dl-3-methyl-2-butanol, 3-methyl-butan-2-ol
PubChem CID 11732
ChEBI CHEBI:77517
IUPAC Name 3-methylbutan-2-ol
SMILES CC(C)C(C)O

Specifications

Density 0.818
Boiling Point 112°C
Flash Point 26°C (78°F)
Refractive Index 1.409
Quantity 100 g
UN Number UN1105
Beilstein 1718800
Merck Index 14,6031
Solubility Information Miscible in Ether,Alcohols. Slightly soluble in water. (2.8g/100mL,30°C)
Formula Weight 88.15
Percent Purity 98%
Chemical Name or Material (+/-)-3-Methyl-2-butanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.