missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1H-pyrazol-5-amine, 95%, Maybridge

Product Code. 10092343
Change view
Click to view available options
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10092343 Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10092343 Supplier Thermo Scientific Maybridge Supplier No. KM00084DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 31230-17-8
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C4H7N3
MDL Number MFCD00051640
Quantity 1 g
Synonym 3-amino-5-methylpyrazole, 3-methyl-1h-pyrazol-5-amine, 1h-pyrazol-3-amine, 5-methyl, 5-amino-3-methylpyrazole, 3-amino-5-methyl-1h-pyrazole, 5-amino-3-methyl-1h-pyrazole, 5-methyl-2h-pyrazol-3-ylamine, 5-methyl-2h-pyrazol-3-amine, 5-methyl-1h-pyrazol-3-ylamine, 5-methylpyrazol-3-amine
InChI Key FYTLHYRDGXRYEY-UHFFFAOYSA-N
SMILES CC1=CC(=NN1)N
IUPAC Name 5-methyl-1H-pyrazol-3-amine
Molecular Weight (g/mol) 97.121
PubChem CID 93146
Formula Weight 97.12
Percent Purity 95%
Chemical Name or Material 3-Methyl-1H-pyrazol-5-amine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.