missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-phenyl-2-pyrazolin-5-one, 99%

Product Code. 10755221 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
This item is not returnable. View return policy

Product Code. 10755221

Brand: Thermo Scientific Acros 127465000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 89-25-8 | C10H10N2O | 174.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-25-8
Molecular Formula C10H10N2O
Molecular Weight (g/mol) 174.2
MDL Number MFCD00003138
InChI Key QELUYTUMUWHWMC-UHFFFAOYSA-N
Synonym edaravone, 3-methyl-1-phenyl-2-pyrazolin-5-one, norphenazone, radicut, 1-phenyl-3-methyl-5-pyrazolone, developer z, methylphenylpyrazolone, norantipyrine, c.i. developer 1, phenylmethylpyrazolone
PubChem CID 4021
ChEBI CHEBI:31530
IUPAC Name 5-methyl-2-phenyl-4H-pyrazol-3-one
SMILES CC1=NN(C(=O)C1)C2=CC=CC=C2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 127.0°C to 131.0°C
Color Beige to Yellow
Boiling Point 287.0°C (265.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 500 g
Beilstein 24, 20
Merck Index 15, 6802
Solubility Information Solubility in water: 3g/L water (20°C). Other solubilities: slightly soluble in alcohol and benzene,insoluble in ether,soluble in acids
Formula Weight 174.2
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 3-Methyl-1-phenyl-2-pyrazolin-5-one, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methyl-1-phenyl-2-pyrazolin-5-one
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.