missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-phenyl-2-pyrazolin-5-one, 98+%

Product Code. 11419093
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419093 2500 g 2500g
11499083 100 g 100g
11409093 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11419093 Supplier Thermo Scientific Alfa Aesar Supplier No. A11161.0E
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 89-25-8 | C10H10N2O | 174.203 g/mol

3-Methyl-1-phenyl-2-pyrazolin-5-one used as reagent for detection of reducing carbohydrates by ESI/MALDI -MS.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-1-phenyl-2-pyrazolin-5-one used as reagent for detection of reducing carbohydrates by ESI/MALDI -MS.

Solubility
Soluble in water(3.3g/L).

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-25-8
Molecular Formula C10H10N2O
Molecular Weight (g/mol) 174.203
MDL Number MFCD00003138
InChI Key QELUYTUMUWHWMC-UHFFFAOYSA-N
Synonym edaravone, 3-methyl-1-phenyl-2-pyrazolin-5-one, norphenazone, radicut, 1-phenyl-3-methyl-5-pyrazolone, developer z, methylphenylpyrazolone, norantipyrine, c.i. developer 1, phenylmethylpyrazolone
PubChem CID 4021
ChEBI CHEBI:31530
IUPAC Name 5-methyl-2-phenyl-4H-pyrazol-3-one
SMILES CC1=NN(C(=O)C1)C2=CC=CC=C2

Specifications

Melting Point 127°C to 131°C
Density 1.12
Boiling Point 191°C (17 mmHg)
Odor Odorless
Quantity 2500 g
Beilstein 609575
Merck Index 14,6713
Solubility Information Soluble in water(3.3g/L).
Formula Weight 174.2
Percent Purity ≥98%
Chemical Name or Material 3-Methyl-1-phenyl-2-pyrazolin-5-one
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.