missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-phenyl-2-phospholene 1-oxide, 94%

Product Code. 11479373
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479373 25 g 25g
11469373 5 g 5g
11489373 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479373 Supplier Thermo Scientific Alfa Aesar Supplier No. A11792.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 707-61-9 | C11H13OP | 192.198 g/mol

3-Methyl-1-phenyl-2-phospholene 1-oxide is used as a catalyst for intramolecular aza-wittig cyclization reactions and polymerization reactions. It is a reactant used for the preparation of phospha sugars and their radical bromination derivatives as anticancer agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-1-phenyl-2-phospholene 1-oxide is used as a catalyst for intramolecular aza-wittig cyclization reactions and polymerization reactions. It is a reactant used for the preparation of phospha sugars and their radical bromination derivatives as anticancer agents.

Solubility
Soluble in polar solvents.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 707-61-9
Molecular Formula C11H13OP
Molecular Weight (g/mol) 192.198
MDL Number MFCD00014518
InChI Key YMKWWHFRGALXLE-UHFFFAOYSA-N
Synonym 3-methyl-1-phenyl-2-phospholene 1-oxide, 3-methyl-1-phenyl-2-phospholene-1-oxide, 2,3-dihydro-4-methyl-1-phenyl-1h-phosphole 1-oxide, 1h-phosphole, 2,3-dihydro-4-methyl-1-phenyl-, 1-oxide, 4-methyl-1-phenyl-2,3-dihydro-1h-phosphole 1-oxide, 2-phospholene, 3-methyl-1-phenyl-, 1-oxide, 2,3-dihydro-4-methyl-1-phenyl-1h-phosphole, 1-oxide, 4-methyl-1-phenyl-2,3-dihydro-1$l^ 5-phosphole 1-oxide, 3-methyl-1-phenyl-4,5-dihydro-1??-phosphol-1-one, 2-phospholene, 1-oxide
PubChem CID 69722
IUPAC Name 4-methyl-1-phenyl-2,3-dihydro-1$l^{5}-phosphole 1-oxide
SMILES CC1=CP(=O)(CC1)C2=CC=CC=C2

Specifications

Melting Point 60°C to 65°C
Boiling Point 173°C to 174°C (0.7 mmHg)
Odor Unpleasant
Quantity 25 g
Beilstein 609223
Sensitivity Hygroscopic
Solubility Information Soluble in polar solvents.
Formula Weight 192.19
Percent Purity 94%
Chemical Name or Material 3-Methyl-1-phenyl-2-phospholene 1-oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.