missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-pentanol, 99+%, Thermo Scientific Chemicals

Product Code. 10307851
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 589-35-5
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
MDL Number MFCD00002937
InChI Key IWTBVKIGCDZRPL-UHFFFAOYSA-N
Synonym 3-methyl-1-pentanol, 3-methylpentanol, 1-pentanol, 3-methyl, 2-ethyl-4-butanol, 3-ethyl-1-butanol, 3-ethylbutanol, 3-methyl pentanol, fema no. 3762, 3-methyl pentan-1-ol, 3-methyl-pentan-1-ol
PubChem CID 11508
ChEBI CHEBI:87381
IUPAC Name 3-methylpentan-1-ol
SMILES CCC(C)CCO

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color Colorless
Density 0.8230g/mL
Boiling Point 150°C to 154°C
Flash Point 58°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH(CH3)CH2CH2OH
Refractive Index 1.4190 to 1.4210 (20°C, 589nm)
Quantity 5 g
Specific Gravity 0.823
Solubility Information Solubility in water: insoluble
Formula Weight 102.18
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 3-Methyl-1-pentanol
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methyl-1-pentanol
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'