missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-1-pentanol, 99+%, Thermo Scientific Chemicals
£23.40 - £182.00
Chemical Identifiers
CAS | 589-35-5 |
---|---|
Molecular Formula | C6H14O |
Molecular Weight (g/mol) | 102.18 |
MDL Number | MFCD00002937 |
InChI Key | IWTBVKIGCDZRPL-UHFFFAOYSA-N |
Synonym | 3-methyl-1-pentanol, 3-methylpentanol, 1-pentanol, 3-methyl, 2-ethyl-4-butanol, 3-ethyl-1-butanol, 3-ethylbutanol, 3-methyl pentanol, fema no. 3762, 3-methyl pentan-1-ol, 3-methyl-pentan-1-ol |
PubChem CID | 11508 |
ChEBI | CHEBI:87381 |
IUPAC Name | 3-methylpentan-1-ol |
SMILES | CCC(C)CCO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10307851
|
Thermo Scientific Acros
335320050 |
5 g | Glass bottle |
£23.40
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10031744
|
Thermo Scientific Acros
335320250 |
25 g | Glass bottle |
£63.60
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10755905
|
Thermo Scientific Acros
335321000 |
100 g | Glass bottle |
£182.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
589-35-5 | |
102.18 | |
IWTBVKIGCDZRPL-UHFFFAOYSA-N | |
11508 | |
3-methylpentan-1-ol |
C6H14O | |
MFCD00002937 | |
3-methyl-1-pentanol, 3-methylpentanol, 1-pentanol, 3-methyl, 2-ethyl-4-butanol, 3-ethyl-1-butanol, 3-ethylbutanol, 3-methyl pentanol, fema no. 3762, 3-methyl pentan-1-ol, 3-methyl-pentan-1-ol | |
CHEBI:87381 | |
CCC(C)CCO |
Specifications
589-35-5 | |
100.0 | |
0.8230g/mL | |
58°C | |
99% min. (GC) | |
CH3CH2CH(CH3)CH2CH2OH | |
MFCD00002937 | |
3-methyl-1-pentanol, 3-methylpentanol, 1-pentanol, 3-methyl, 2-ethyl-4-butanol, 3-ethyl-1-butanol, 3-ethylbutanol, 3-methyl pentanol, fema no. 3762, 3-methyl pentan-1-ol, 3-methyl-pentan-1-ol | |
IWTBVKIGCDZRPL-UHFFFAOYSA-N | |
3-methylpentan-1-ol | |
11508 | |
102.18 | |
Liquid |
99.0 | |
Colorless | |
150°C to 154°C | |
Authentic | |
C6H14O | |
1.4190 to 1.4210 (20°C, 589nm) | |
0.823 | |
Solubility in water: insoluble | |
CCC(C)CCO | |
102.18 | |
CHEBI:87381 | |
99+% | |
3-Methyl-1-pentanol |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 209-644-9
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 3-Methyl-1-pentanol
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
RUO – Research Use Only