missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-cyclohexene, 90%, Tech.

Product Code. 10297220 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
This item is not returnable. View return policy

Product Code. 10297220

Brand: Thermo Scientific Acros 304310250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 591-48-0 | C7H12 | 96.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 591-48-0
CAS Min % 88.0
CAS Max % 100.0
Color Colorless
Density 0.8000g/mL
Boiling Point 104°C
Flash Point -3°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Molecular Formula C7H12
Linear Formula C6H9CH3
Refractive Index 1.4425 to 1.4445
MDL Number MFCD00001571
Quantity 25 mL
Beilstein 05, 67
Specific Gravity 0.8
Synonym 3-methyl-1-cyclohexene, cyclohexene, 3-methyl, 3-methyl cyclohexene, 3-methyl-cyclohexene, 3-methylcyclohexene-1, 3-methylcyclohex-1-ene, acmc-1atr1
InChI Key UZPWKTCMUADILM-UHFFFAOYSA-N
SMILES CC1CCCC=C1
IUPAC Name 3-methylcyclohexene
Molecular Weight (g/mol) 96.17
PubChem CID 11573
Formula Weight 96.17
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material 3-Methyl-1-cyclohexene, tech., 90%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methyl-1-cyclohexene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.