missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-butanol, mixture of isomers, 99%

Product Code. 11383187
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383187 500 mL 500mL
11373187 100 mL 100mL
11393187 2500 mL 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11383187 Supplier Thermo Scientific Alfa Aesar Supplier No. L13660.AP
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 123-51-3 | C5H12O | 88.15 g/mol

3-Methyl-1-butanol is used as an active ingredient in banana oil production and Kovac's reagent. It acts as an antifoaming agent. It serves as an intermediate for the manufacture of corrosion inhibitors, flotation aids, fuel and lubricating oil additives, polyvinyl chloride stabilizers, pharmaceuticals, cosmetics, plasticizers, rubber auxiliaries and paint solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-1-butanol is used as an active ingredient in banana oil production and Kovac′s reagent. It acts as an antifoaming agent. It serves as an intermediate for the manufacture of corrosion inhibitors, flotation aids, fuel and lubricating oil additives, polyvinyl chloride stabilizers, pharmaceuticals, cosmetics, plasticizers, rubber auxiliaries and paint solvents.

Solubility
Miscible with water, alcohol, ether, benzene, chloroform, petroleum ether, glacial acetic acid, acetone and oils.

Notes
Incompatible with strong oxidizing agents, strong acids, acid chlorides and acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-51-3
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00002934
InChI Key PHTQWCKDNZKARW-UHFFFAOYSA-N
Synonym 3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol
PubChem CID 31260
ChEBI CHEBI:15837
IUPAC Name 3-methylbutan-1-ol
SMILES CC(C)CCO

Specifications

Melting Point -117°C
Density 0.809
Boiling Point 130°C to 132°C
Flash Point 45°C (113°F)
Odor Unpleasant
Refractive Index 1.406
Quantity 500 mL
UN Number UN1105
Beilstein 1718835
Merck Index 14,5195
Solubility Information Miscible with water,alcohol,ether,benzene,chloroform,petroleum ether,glacial acetic acid,acetone and oils.
Formula Weight 88.15
Percent Purity 99%
Chemical Name or Material 3-Methyl-1-butanol, mixture of isomers
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Methyl-1-butanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 3-Methyl-1-butanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.