missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-butanol, ACS Reagent, ≥98.5%, Honeywell Riedel-de Haën™

Product Code. 15683620
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15683620 500 mL Glass Bottle 500mL
15663620 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15683620 Supplier Honeywell-Riedel-de Haen Supplier No. 320021500ML
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent,≥98.5%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-51-3
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00002934
InChI Key PHTQWCKDNZKARW-UHFFFAOYSA-N
Synonym 3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol
PubChem CID 31260
ChEBI CHEBI:15837
IUPAC Name 3-methylbutan-1-ol
SMILES CC(C)CCO

Specifications

Melting Point -117°C
Density 0.809g/mL at 25°C
Boiling Point 130°C
Explosion Limit 1.2 to 9% (100°F)
Flash Point 43°C (109.4°F)
Ignition Point 644°F
Packaging Glass Bottle
Linear Formula (CH3)2CHCH2CH2OH
Quantity 500 mL
UN Number UN1105
Beilstein 1718835
Vapor Density 3 (vs air)
Formula Weight 88.15g/mol
Vapor Pressure 2mmHg (20°C)
Percent Purity ≥98.5%
Grade ACS Reagent
Chemical Name or Material 3-Methyl-1-Butanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.