missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-Butanol, 98%, Reagent Grade, Honeywell™

Product Code. 15677940 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
100mL
2.5 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15677940 100 mL Glass bottle 100mL
15607950 500 mL Glass bottle 500mL
15687940 1 L Glass bottle 1 L
15697940 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15677940 Supplier Honeywell Research Chemicals Supplier No. M32658100ML
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Denne vare kan ikke returneres. Se returpolicy

Reagent Grade, 98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 123-51-3
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00002934
InChI Key PHTQWCKDNZKARW-UHFFFAOYSA-N
Synonym 3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol
PubChem CID 31260
ChEBI CHEBI:15837
IUPAC Name 3-methylbutan-1-ol
SMILES CC(C)CCO

Tekniske data

Melting Point -117°C
Color Colorless
Density 0.809g/mL at 25°C
Boiling Point 131°C to 132°C
Explosion Limit 1.2 to 9% (100°F)
Flash Point 43°C (109.4°F)
Ignition Point 644°F
Packaging Glass bottle
Linear Formula (CH3)2CHCH2CH2OH
Quantity 100 mL
UN Number UN1105
Beilstein 1718835
Vapor Density 3 (vs air)
Formula Weight 88.15g/mol
Vapor Pressure 2mmHg (20°C)
Percent Purity 0.98
Chemical Name or Material 3-Methyl-1-butanol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.