missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1,5-pentanediol, TRC

Product Code. 30465123
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
This item is not returnable. View return policy

Product Code. 30465123

Brand: Toronto Research Chemicals TRCM49313050ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Methyl-1,5-pentanediol
CAS 4457-71-0
Molecular Formula C6 H14 O2
Quantity 50 mL
Synonym 3-Methyl-1,5-pentanediol, 1,5-Dihydroxy-3-methylpentane, Diol MPD, MPD, NSC 6496, 1,5-Pentanediol, 3-methyl-
InChI Formula InChI=1S/C6H14O2/c1-6(2-4-7)3-5-8/h6-8H,2-5H2,1H3
SMILES CC(CCO)CCO
IUPAC Name 3-methylpentane-1,5-diol
Molecular Weight (g/mol) 118.1742
Formula Weight 118.0994
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.