missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-(2-nitro-phenyl)-piperazine, Thermo Scientific™

Product Code. 10391370
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10391370 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10391370 Supplier Thermo Scientific Maybridge Supplier No. MAY00087DA
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 398470-53-6
Packaging Amber Glass Bottle
Molecular Formula C11H15N3O2
MDL Number MFCD00448002
Quantity 1 g
Synonym 3-methyl-1-2-nitrophenyl piperazine, 3-methyl-1-2-nitro-phenyl-piperazine, 3-methyl-1-2-nitro-phenyl piperazine, acmc-1aien, 1-2-nitrophenyl-3-methylpiperazine, piperazine,3-methyl-1-2-nitrophenyl
InChI Key JLCKZYKOWUVIPC-UHFFFAOYNA-N
SMILES CC1CN(CCN1)C1=CC=CC=C1[N+]([O-])=O
IUPAC Name 3-methyl-1-(2-nitrophenyl)piperazine
Molecular Weight (g/mol) 221.26
PubChem CID 2833123
Formula Weight 221.26
Percent Purity 95%
Chemical Name or Material 3-methyl-1-(2-nitro-phenyl)-piperazine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.