missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1,2-cyclopentanedione, 99%

Product Code. 10022270 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10022270 50 g 50g
10387450 10 g 10g
10720955 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10022270 Supplier Thermo Scientific Acros Supplier No. 176460500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 765-70-8 | C6H8O2 | 112.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 765-70-8
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.13
MDL Number MFCD00001417
InChI Key OACYKCIZDVVNJL-UHFFFAOYSA-N
Synonym 3-methyl-1,2-cyclopentanedione, 1,2-cyclopentanedione, 3-methyl, methylcyclopentenolone diketo form, fema no. 2700, methylcyclopentenolone natural, kentonarome, benzil-related compound, 45, 3-methyl-1,2-cyclopentanedione maple lactone, maple lactone mcp, methyl-cyclopentenolone
PubChem CID 61209
IUPAC Name 3-methylcyclopentane-1,2-dione
SMILES CC1CCC(=O)C1=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 105.0°C to 110.0°C
Color White
Density 100g/mL
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 50 g
Beilstein 07, I, 310
Solubility Information Solubility in water: soluble.
Formula Weight 112.13
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 3-Methyl-1,2-cyclopentanedione
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.