missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-1,2-benzenediol, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 3-Methylcatechol |
| CAS | 488-17-5 |
| Molecular Formula | C7 H8 O2 |
| Quantity | 1 g |
| Synonym | 1,2-Dihydroxy-3-methylbenzene, 2,3-Dihydroxytoluene, 2,3-Toulenediol, 3-Methyl-1,2-benzenediol, 3-Methylpyrocatechol |
| InChI Formula | InChI=1S/C7H8O2/c1-5-3-2-4-6(8)7(5)9/h2-4,8-9H,1H3 |
| SMILES | Cc1cccc(O)c1O |
| IUPAC Name | 3-methylbenzene-1,2-diol |
| Molecular Weight (g/mol) | 124.14 |
| Formula Weight | 124.0524 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction