missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxytyramine Antibody, Novus Biologicals™

Product Code. 18386826 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18386826 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18386826 Supplier Novus Biologicals Supplier No. NBP313405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

3-Methoxytyramine Polyclonal antibody specifically detects 3-Methoxytyramine in All species samples. It is validated for ELISA, Dot Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen 3-Methoxytyramine
Applications ELISA, Dot Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 1% BSA, 20% Glycerol (pH7)
Gene Alias 3-methoxy-4-hydroxyphenethylamine, 3MT, 3-MT, 3-O-Methyldopamine, 4-(2-Aminoethyl)-2-methoxyphenol
Host Species Rabbit
Immunogen The immunogen used to generate this antibody corresponds to 3-Methoxytyramine
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species All species
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.