missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxythiophenol, 97%

Product Code. 11483384
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11483384 5 g 5g
11369469 1 g 1g
11493384 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11483384 Supplier Thermo Scientific Alfa Aesar Supplier No. A10648.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15570-12-4 | C7H8OS | 140.2 g/mol

3-Methoxythiophenol is used in the preparation of allenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxythiophenol is used in the preparation of allenes.

Solubility
Not miscible or difficult to mix in water. Soluble in methanol, benzene, hexane, toluene and dichloromethane.

Notes
Store under dry inert gas. It is sensitive to air. Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15570-12-4
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.2
MDL Number MFCD00004841
InChI Key QMVAZEHZOPDGHA-UHFFFAOYSA-N
Synonym 3-methoxythiophenol, 3-methoxybenzenthiol, 3-methoxy thiophenol, m-methoxythiophenol, 3-mercaptoanisole, benzenethiol, 3-methoxy, m-methoxybenzenethiol, 3-methoxy-benzenethiol, 3-methoxybenzene-1-thiol, meta-methoxybenzenethiol
PubChem CID 84989
IUPAC Name 3-methoxybenzenethiol
SMILES COC1=CC(=CC=C1)S

Specifications

Density 1.136
Boiling Point 225°C to 227°C
Flash Point 96°C (204°F)
Odor Unpleasant
Refractive Index 1.5865
Quantity 5 g
Beilstein 2041496
Sensitivity Air Sensitive
Solubility Information Not miscible or difficult to mix in water. Soluble in methanol,benzene,hexane,toluene and dichloromethane.
Formula Weight 140.21
Percent Purity 98%
Chemical Name or Material 3-Methoxythiophenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.