missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxythiophene, 99%

Product Code. 11475037
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11475037 25 g 25g
11455037 1 g 1g
11465037 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11475037 Supplier Thermo Scientific Alfa Aesar Supplier No. B21940.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 17573-92-1 | C5H6OS | 114.162 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17573-92-1
Molecular Formula C5H6OS
Molecular Weight (g/mol) 114.162
MDL Number MFCD00043894
InChI Key RFSKGCVUDQRZSD-UHFFFAOYSA-N
Synonym thiophene, 3-methoxy, 3-methoxy-thiophene, pubchem8863, acmc-1bow6, 3-methoxythiophene, ksc491i4p, bidd:gt0411
PubChem CID 140249
IUPAC Name 3-methoxythiophene
SMILES COC1=CSC=C1

Specifications

Density 1.147
Boiling Point 60°C to 61°C (12 mmHg)
Flash Point 49°C (120°F)
Odor Recognizable
Refractive Index 1.532
Quantity 25 g
UN Number UN3271
Beilstein 106404
Sensitivity Air sensitive
Formula Weight 114.17
Percent Purity 99%
Chemical Name or Material 3-Methoxythiophene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.