missing translation for 'onlineSavingsMsg'
Learn More

3-methoxythiophene-2-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10209643
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10209643 250 mg Amber Glass Bottle 250mg
10452104 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10209643 Supplier Thermo Scientific Maybridge Supplier No. CC68604CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 35134-07-7
Molecular Formula C6H6O2S
Molecular Weight (g/mol) 142.17
MDL Number MFCD01571098
InChI Key KGJDTMQUUPIAEF-UHFFFAOYSA-N
Synonym 3-methoxy-2-thiophenecarboxaldehyde, 3-methoxythiophene-2-carboxaldehyde, 2-thiophenecarboxaldehyde, 3-methoxy, 3-methoxy-2-thenaldehyde, 2-formyl-3-methoxythiophene, 3-methoxy-2-thiophenealdehyde, 3-methoxy-2-thiophenecarbaldehyde, 3-methoxy-thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde,3-methoxy, 3-methoxy-2-thiophenecarbaldehyde #
PubChem CID 588315
IUPAC Name 3-methoxythiophene-2-carbaldehyde
SMILES COC1=C(SC=C1)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 142.18
Percent Purity 97%
Chemical Name or Material 3-Methoxythiophene-2-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.