missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxypropionitrile, 98%

Product Code. 10347181 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10347181 100 mL Glass bottle 100mL
10707592 500 mL Glass bottle 500mL
10197463 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10347181 Supplier Thermo Scientific Acros Supplier No. 257441000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-67-8 | C4H7NO | 85.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-67-8
Molecular Formula C4H7NO
Molecular Weight (g/mol) 85.11
MDL Number MFCD00001958
InChI Key OOWFYDWAMOKVSF-UHFFFAOYSA-N
Synonym 3-methoxypropionitrile, propanenitrile, 3-methoxy, 2-cyanoethyl methyl ether, 3-methoxypropylnitrile, beta-methoxypropionitrile, 1-cyano-2-methoxyethane, propionitrile, 3-methoxy, 3-methoxypropannitril, 3-methoxypropiononitrile, beta-methyoxypropionitrile
PubChem CID 61032
IUPAC Name 3-methoxypropanenitrile
SMILES COCCC#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -62.9°C
Color Colorless to Yellow
Density 0.9380g/mL
Boiling Point 164.0°C to 165.0°C
Flash Point 66°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3OCH2CH2CN
Refractive Index 1.4020 to 1.4040
Quantity 100 mL
Specific Gravity 0.938
Solubility Information Solubility in water: 335g/L (20°C)
Formula Weight 85.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Methoxypropionitrile, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.