missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxypropionic acid, 97+%

Product Code. 15294288 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15294288 50 mL 50mL
15284288 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15294288 Supplier Thermo Scientific Acros Supplier No. 453320500
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2544-06-1 | C4H8O3 | 104.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2544-06-1
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.11
InChI Key YSIKHBWUBSFBRZ-UHFFFAOYSA-N
PubChem CID 134442
IUPAC Name 3-methoxypropanoic acid
SMILES COCCC(=O)O

Specifications

Density 1.1080g/mL
Boiling Point 116.0°C (9.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 50 mL
Specific Gravity 1.108
Synonym 3-methoxypropionic acid, propanoic acid, 3-methoxy, propanoic acid, methoxy, m-peg1-acid, propanoic acid,methoxy, acmc-1aiwk, 3-methoxy-propionic acid, #, propanoic acid,3-methoxy, .beta.-methoxypropionic acid
Formula Weight 104.11
Percent Purity 97+%
Chemical Name or Material 3-Methoxypropionic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methoxypropionic acid, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.