missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxypiperidine hydrochloride, Thermo Scientific™

Product Code. 15468017
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15468017 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15468017 Supplier Thermo Scientific Alfa Aesar Supplier No. H51138.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 688809-94-1
Molecular Formula C6H14ClNO
Molecular Weight (g/mol) 151.634
MDL Number MFCD06800959
InChI Key ZRDDHBSVIQRILE-UHFFFAOYSA-N
Synonym 3-methoxypiperidine hydrochloride, 3-methoxy-piperidine hydrochloride, 3-methoxypiperidine hcl, 3-methoxy piperidine hcl, piperidine, 3-methoxy-, hydrochloride, 3-methoxypiperidine-hydrogen chloride 1/1, 4045-29-8 3-methoxypiperidine hydrochloride
PubChem CID 23142000
IUPAC Name 3-methoxypiperidine;hydrochloride
SMILES COC1CCCNC1.Cl

Specifications

Quantity 1 g
Sensitivity Hygroscopic
Formula Weight 151.64
Chemical Name or Material 3-Methoxypiperidine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.