missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenylacetylene, 96%

Product Code. 15471438
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15471438 250 mg 250mg
15451438 1 g 1g
15461438 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15471438 Supplier Thermo Scientific Alfa Aesar Supplier No. H55254.MD
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 768-70-7 | C9H8O | 132.16 g/mol

It is a useful aromatic compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a useful aromatic compound.

Solubility
Slightly soluble in water.

Notes
Storage: Keep container tightly closed. Store in a cool, dark and well-ventilated place and away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 768-70-7
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.16
MDL Number MFCD00160810
InChI Key ZASXCTCNZKFDTP-UHFFFAOYSA-N
Synonym 3-ethynylanisole, 3-methoxyphenylacetylene, 1-ethynyl-3-methoxy-benzene, benzene, 1-ethynyl-3-methoxy, 3-ethynyl-1-methoxybenzene, 1-etynyl-3-methoxy-benzene, 3-ethynyl-anisole, m-methoxyphenylacetylene, 3-ethynylanisole 96
PubChem CID 640753
IUPAC Name 1-ethynyl-3-methoxybenzene
SMILES COC1=CC=CC(=C1)C#C

Specifications

Density 1.04
Boiling Point 204°C to 210°C
Flash Point 76°C (169°F)
Refractive Index 1.555
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 132.16
Percent Purity 96%
Chemical Name or Material 3-Methoxyphenylacetylene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.