missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenylacetone, 97%

Product Code. 11429956
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429956 5 g 5g
11419956 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11429956 Supplier Thermo Scientific Alfa Aesar Supplier No. B20916.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3027-13-2 | C10H12O2 | 164.20 g/mol

3-Methoxyphenylacetone is employed in the preparation of optically active cyanohydrins. It is also used in the synthesis of central nervous system (CNS) active compounds. Further, it plays an important role in the preparation of 4-amino-3-methoxypropiophenone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxyphenylacetone is employed in the preparation of optically active cyanohydrins. It is also used in the synthesis of central nervous system (CNS) active compounds. Further, it plays an important role in the preparation of 4-amino-3-methoxypropiophenone.

Solubility
Miscible with dimethyl sulfoxide.

Notes
Incompatible with strong oxidizing agents, strong acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3027-13-2
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00008771
InChI Key RMMRRRLPDBJBQL-UHFFFAOYSA-N
Synonym 3-methoxyphenylacetone, 1-3-methoxyphenyl propan-2-one, 1-3-methoxyphenyl acetone, 3-methoxyphenyl acetone, m-methoxyphenyl acetone, m-methoxyphenylacetone, 2-propanone, 1-3-methoxyphenyl, 3-methoxyphenyl methyl ketone, acmc-1ai0k
PubChem CID 76410
IUPAC Name 1-(3-methoxyphenyl)propan-2-one
SMILES COC1=CC=CC(CC(C)=O)=C1

Specifications

Density 1.078
Boiling Point 258°C to 260°C
Flash Point >110°C (230°F)
Refractive Index 1.5252
Quantity 5 g
Beilstein 2044357
Solubility Information Miscible with dimethyl sulfoxide.
Formula Weight 164.2
Percent Purity 97%
Chemical Name or Material 3-Methoxyphenylacetone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.