missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenylacetic acid, 99.5%

Product Code. 10703475 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10703475 100 g Glass bottle 100g
10091280 5 g Glass bottle 5g
10645942 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10703475 Supplier Thermo Scientific Acros Supplier No. 126011000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1798-09-0 | C9H10O3 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1798-09-0
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00004334
InChI Key LEGPZHPSIPPYIO-UHFFFAOYSA-N
Synonym 3-methoxyphenylacetic acid, 2-3-methoxyphenyl acetic acid, m-methoxyphenylacetic acid, 3-methoxyphenyl acetic acid, 3-methoxybenzeneacetic acid, benzeneacetic acid, 3-methoxy, acetic acid, m-methoxyphenyl, unii-25xlo0t6my, 3-methoxy-phenyl-acetic acid, 25xlo0t6my
PubChem CID 15719
IUPAC Name 2-(3-methoxyphenyl)acetic acid
SMILES COC1=CC=CC(=C1)CC(=O)O

Specifications

CAS Min % 99.4
CAS Max % 100.0
Melting Point 71.0°C to 73.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99.50%
Packaging Glass bottle
Linear Formula CH3OC6H4CH2CO2H
Quantity 100 g
Beilstein 10, I, 82
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol
Formula Weight 166.18
Percent Purity 99.5%
Physical Form Flakes
Chemical Name or Material 3-Methoxyphenylacetic acid
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • 3-Methoxyphenylacetic acid
Signalwort
  • Warning
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.