missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenylacetic acid, 97%

Product Code. 11404383
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404383 100 g 100g
11484373 5 g 5g
11494373 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11404383 Supplier Thermo Scientific Alfa Aesar Supplier No. A10951.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1798-09-0 | C9H10O3 | 166.176 g/mol

A fluorimetric method for the estimation of 4-hydroxy-3-methoxyphenylacetic acid (homovanillic acid) has been developed and applied to normal brain tissue. The presence of homovanillic acid in the caudate nucleus of normal animals of several species has been demonstrated.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A fluorimetric method for the estimation of 4-hydroxy-3-methoxyphenylacetic acid (homovanillic acid) has been developed and applied to normal brain tissue. The presence of homovanillic acid in the caudate nucleus of normal animals of several species has been demonstrated.

Solubility
Soluble in Chloroform and Ethyl Acetate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1798-09-0
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00004334
InChI Key LEGPZHPSIPPYIO-UHFFFAOYSA-N
Synonym 3-methoxyphenylacetic acid, 2-3-methoxyphenyl acetic acid, m-methoxyphenylacetic acid, 3-methoxyphenyl acetic acid, 3-methoxybenzeneacetic acid, benzeneacetic acid, 3-methoxy, acetic acid, m-methoxyphenyl, unii-25xlo0t6my, 3-methoxy-phenyl-acetic acid, 25xlo0t6my
PubChem CID 15719
IUPAC Name 2-(3-methoxyphenyl)acetic acid
SMILES COC1=CC=CC(=C1)CC(=O)O

Specifications

Melting Point 67°C to 72°C
Quantity 100 g
Beilstein 2614004
Solubility Information Soluble in Chloroform and Ethyl Acetate.
Formula Weight 166.18
Percent Purity 97%
Chemical Name or Material 3-Methoxyphenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.