missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methoxyphenylacetic acid, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
A fluorimetric method for the estimation of 4-hydroxy-3-methoxyphenylacetic acid (homovanillic acid) has been developed and applied to normal brain tissue. The presence of homovanillic acid in the caudate nucleus of normal animals of several species has been demonstrated.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A fluorimetric method for the estimation of 4-hydroxy-3-methoxyphenylacetic acid (homovanillic acid) has been developed and applied to normal brain tissue. The presence of homovanillic acid in the caudate nucleus of normal animals of several species has been demonstrated.
Solubility
Soluble in Chloroform and Ethyl Acetate.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
A fluorimetric method for the estimation of 4-hydroxy-3-methoxyphenylacetic acid (homovanillic acid) has been developed and applied to normal brain tissue. The presence of homovanillic acid in the caudate nucleus of normal animals of several species has been demonstrated.
Solubility
Soluble in Chloroform and Ethyl Acetate.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Melting Point | 67°C to 72°C |
Quantity | 25 g |
Beilstein | 2614004 |
Solubility Information | Soluble in Chloroform and Ethyl Acetate. |
Formula Weight | 166.18 |
Percent Purity | 97% |
Chemical Name or Material | 3-Methoxyphenylacetic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'